Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATTAAACTTCTGTCATCCATGA[C/T]AGATGCTAATAAATTCCAGGTGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASTKD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FASTKD3 - FAST kinase domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024091.3 | Intron | NP_076996.2 | ||||
XM_006714498.1 | Intron | XP_006714561.1 |
MTRR - 5-methyltetrahydrofolate-homocysteine methyltransferase reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002454.2 | Intron | NP_002445.2 | ||||
NM_024010.2 | Intron | NP_076915.2 |