Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCTTCCCCTATCGAGCGTACACC[A/G]ATGGAGGAGGTATGGGCCTGAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606205 | ||||||||||||||||||||
Literature Links: |
SLC6A7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC6A7 - solute carrier family 6 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014228.4 | 471 | Missense Mutation | AAT,GAT | N,D 70 | NP_055043.2 | |
XM_017009767.1 | 471 | Missense Mutation | AAT,GAT | N,D 151 | XP_016865256.1 | |
XM_017009768.1 | 471 | UTR 5 | XP_016865257.1 | |||
XM_017009769.1 | 471 | UTR 5 | XP_016865258.1 | |||
XM_017009770.1 | 471 | UTR 5 | XP_016865259.1 |