Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCCCTTGCCTCCATTGCAGCC[A/G]CAGCAAGAGGCCTCCACTTGTCCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605166 MIM: 611213 | ||||||||||||||||||||
Literature Links: |
FCHSD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCHSD1 - FCH and double SH3 domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033449.2 | 1278 | UTR 3 | NP_258260.1 | |||
XM_005268524.4 | 1278 | Intron | XP_005268581.1 | |||
XM_006714803.3 | 1278 | Intron | XP_006714866.1 | |||
XM_011537698.2 | 1278 | Intron | XP_011536000.1 | |||
XM_011537700.2 | 1278 | Intron | XP_011536002.1 | |||
XM_011537701.2 | 1278 | Intron | XP_011536003.1 | |||
XM_017010013.1 | 1278 | Intron | XP_016865502.1 |
HDAC3 - histone deacetylase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RELL2 - RELT like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130029.1 | 1278 | Missense Mutation | CAC,CGC | H,R 137 | NP_001123501.1 | |
NM_173828.4 | 1278 | Missense Mutation | CAC,CGC | H,R 137 | NP_776189.3 | |
XM_005268414.3 | 1278 | Missense Mutation | CAC,CGC | H,R 137 | XP_005268471.1 | |
XM_011537624.1 | 1278 | Missense Mutation | CAC,CGC | H,R 137 | XP_011535926.1 | |
XM_011537625.2 | 1278 | Missense Mutation | CAC,CGC | H,R 137 | XP_011535927.1 |