Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCGATTTCTTCGAGGTGGAGTC[A/G]GAGCTGGGACGGTAAGGCGCGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMK4 - calcium/calmodulin dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323374.1 | 249 | Silent Mutation | TCA,TCG | S,S 50 | NP_001310303.1 | |
NM_001323375.1 | 249 | Silent Mutation | TCA,TCG | S,S 50 | NP_001310304.1 | |
NM_001323376.1 | 249 | UTR 5 | NP_001310305.1 | |||
NM_001323377.1 | 249 | UTR 5 | NP_001310306.1 | |||
NM_001744.5 | 249 | Silent Mutation | TCA,TCG | S,S 50 | NP_001735.1 |