Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCACATCTCTGTGTAGATCAAGC[A/G]TATCACCTCTGTAAAGATGACTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610499 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAPGEF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAPGEF6 - Rap guanine nucleotide exchange factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164386.1 | 5575 | UTR 3 | NP_001157858.1 | |||
NM_001164387.1 | 5575 | UTR 3 | NP_001157859.1 | |||
NM_001164388.1 | 5575 | UTR 3 | NP_001157860.1 | |||
NM_001164389.1 | 5575 | Intron | NP_001157861.1 | |||
NM_001164390.1 | 5575 | Intron | NP_001157862.1 | |||
NM_016340.5 | 5575 | UTR 3 | NP_057424.3 |