Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAAAGGCCCACATAAATGGATA[C/T]GGCAACAAGACCAAATTGCAGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607124 | ||||||||||||||||||||
Literature Links: |
PLPP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLPP1 - phospholipid phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003711.3 | 1214 | Missense Mutation | ATA,GTA | I,V 209 | NP_003702.2 | |
NM_176895.2 | 1214 | Missense Mutation | ATA,GTA | I,V 210 | NP_795714.1 | |
XM_006714724.2 | 1214 | Missense Mutation | ATA,GTA | I,V 146 | XP_006714787.1 |
SKIV2L2 - Ski2 like RNA helicase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015360.4 | 1214 | Intron | NP_056175.3 |