Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGGTTGTTGATCTTGTTCTTCTC[C/T]ACTTGTTCACTCTGGCGCTGGTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118970 MIM: 612861 | ||||||||||||||||||||
Literature Links: |
ARL10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL10 - ADP ribosylation factor like GTPase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317948.1 | 646 | Intron | NP_001304877.1 | |||
NM_173664.5 | 646 | Intron | NP_775935.1 | |||
XM_011534529.2 | 646 | Intron | XP_011532831.1 | |||
XM_011534530.2 | 646 | Intron | XP_011532832.1 | |||
XM_011534531.2 | 646 | Intron | XP_011532833.1 | |||
XM_017009372.1 | 646 | Intron | XP_016864861.1 |
CLTB - clathrin light chain B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001834.3 | 646 | Silent Mutation | GTA,GTG | V,V 147 | NP_001825.1 | |
NM_007097.3 | 646 | Silent Mutation | GTA,GTG | V,V 147 | NP_009028.1 | |
XM_017009026.1 | 646 | Silent Mutation | GTA,GTG | V,V 80 | XP_016864515.1 | |
XM_017009027.1 | 646 | Intron | XP_016864516.1 |
HIGD2A - HIG1 hypoxia inducible domain family member 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOP16 - NOP16 nucleolar protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |