Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGGACCCACCACTGCCAAAATC[C/T]CCTGAGGTTTCCTGCTCCTCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPOCK1 - sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004598.3 | 1303 | Silent Mutation | GGA,GGG | G,G 384 | NP_004589.1 |