Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACTGTGATGGAAGAACTAGAGG[A/T]TATTGTGATCGCGTATGGACAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
THG1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THG1L - tRNA-histidine guanylyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317824.1 | 339 | UTR 5 | NP_001304753.1 | |||
NM_001317825.1 | 339 | UTR 5 | NP_001304754.1 | |||
NM_001317826.1 | 339 | Missense Mutation | GAT,GTT | D,V 34 | NP_001304755.1 | |
NM_017872.4 | 339 | Missense Mutation | GAT,GTT | D,V 96 | NP_060342.2 | |
XM_017009603.1 | 339 | Intron | XP_016865092.1 |