Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGAGGTGGCCATACCGTTGAC[C/T]GCTCCTCCAACTAACCAGTAAGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASA1 - RAS p21 protein activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002890.2 | 1173 | Silent Mutation | ACC,ACT | T,T 174 | NP_002881.1 | |
NM_022650.2 | 1173 | UTR 5 | NP_072179.1 | |||
XM_011543525.1 | 1173 | Silent Mutation | ACC,ACT | T,T 174 | XP_011541827.1 | |
XM_011543527.2 | 1173 | Silent Mutation | ACC,ACT | T,T 174 | XP_011541829.1 |