Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGCTATAGGGGATAATCTGTT[C/T]TCTTTTTCTTTTCTTAGCTTCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf63 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf63 - chromosome 5 open reading frame 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164478.1 | 2895 | Intron | NP_001157950.1 | |||
NM_001164479.1 | 2895 | UTR 3 | NP_001157951.1 | |||
XM_017009464.1 | 2895 | Intron | XP_016864953.1 | |||
XM_017009465.1 | 2895 | Intron | XP_016864954.1 |