Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCTGGGCCCCTACGCCTCTGG[C/T]TCATACTCCTGATATTCCTCCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602569 | ||||||||||||||||||||
Literature Links: |
EIF4E1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF4E1B - eukaryotic translation initiation factor 4E family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4281 - microRNA 4281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNCB - synuclein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001502.2 | 684 | Silent Mutation | GAA,GAG | E,E 131 | NP_001001502.1 | |
NM_001318034.1 | 684 | Silent Mutation | GAA,GAG | E,E 117 | NP_001304963.1 | |
NM_001318035.1 | 684 | Silent Mutation | GAA,GAG | E,E 101 | NP_001304964.1 | |
NM_001318036.1 | 684 | Silent Mutation | GAA,GAG | E,E 117 | NP_001304965.1 | |
NM_001318037.1 | 684 | Silent Mutation | GAA,GAG | E,E 101 | NP_001304966.1 | |
NM_003085.4 | 684 | Silent Mutation | GAA,GAG | E,E 131 | NP_003076.1 | |
XM_006714914.3 | 684 | Intron | XP_006714977.1 | |||
XM_006714915.3 | 684 | Intron | XP_006714978.1 | |||
XM_006714916.2 | 684 | Silent Mutation | GAA,GAG | E,E 131 | XP_006714979.1 | |
XM_011534640.2 | 684 | Intron | XP_011532942.1 |