Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCCATGAAGCTACATTGTTGC[C/T]TATTCACTTTAGTGGCAAGTATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFAP3 - microfibrillar associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135037.1 | 238 | Intron | NP_001128509.1 | |||
NM_001242336.1 | 238 | Silent Mutation | CTA,TTA | L,L 7 | NP_001229265.1 | |
NM_005927.4 | 238 | Silent Mutation | CTA,TTA | L,L 7 | NP_005918.1 |