Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGTTGCTCATAGGAGACGATAG[C/T]GATATATTTTCTAATAAGAAAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601874 | ||||||||||||||||||||
Literature Links: |
ELL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELL2 - elongation factor for RNA polymerase II 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012081.5 | 1728 | Missense Mutation | ACT,GCT | T,A 534 | NP_036213.2 | |
XM_006714575.2 | 1728 | Missense Mutation | ACT,GCT | T,A 479 | XP_006714638.1 | |
XM_017009239.1 | 1728 | Missense Mutation | ACT,GCT | T,A 534 | XP_016864728.1 | |
XM_017009240.1 | 1728 | Missense Mutation | ACT,GCT | T,A 402 | XP_016864729.1 | |
XM_017009241.1 | 1728 | Missense Mutation | ACT,GCT | T,A 402 | XP_016864730.1 | |
XM_017009242.1 | 1728 | Missense Mutation | ACT,GCT | T,A 402 | XP_016864731.1 | |
XM_017009243.1 | 1728 | Missense Mutation | ACT,GCT | T,A 349 | XP_016864732.1 |