Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAATTTACTTTTTATAAAAAGA[A/C]AAATGTACATATTTACACACGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf56 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf56 - chromosome 5 open reading frame 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRF1 - interferon regulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002198.2 | 1928 | UTR 3 | NP_002189.1 | |||
XM_011543378.2 | 1928 | UTR 3 | XP_011541680.1 | |||
XM_011543379.2 | 1928 | UTR 3 | XP_011541681.1 |