Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATAATAGATCACAAACAGGAAA[G/T]TCCTGGCTATTTTACAGATATAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHD1 - chromodomain helicase DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270.2 | 3635 | UTR 3 | NP_001261.2 | |||
XM_005271866.4 | 3635 | UTR 3 | XP_005271923.1 | |||
XM_005271867.4 | 3635 | UTR 3 | XP_005271924.1 | |||
XM_011543112.2 | 3635 | UTR 3 | XP_011541414.1 | |||
XM_017008991.1 | 3635 | UTR 3 | XP_016864480.1 | |||
XM_017008992.1 | 3635 | UTR 3 | XP_016864481.1 |