Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGACCAGGGGTCTCCTCCCTCT[A/G]ACCAGCCTCCAGTCTGGGGAGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAXDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAXDC2 - fatty acid hydroxylase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032385.4 | 1108 | UTR 3 | NP_115761.2 | |||
XM_005268358.2 | 1108 | UTR 3 | XP_005268415.1 | |||
XM_006714753.2 | 1108 | UTR 3 | XP_006714816.1 | |||
XM_017008965.1 | 1108 | UTR 3 | XP_016864454.1 | |||
XM_017008966.1 | 1108 | UTR 3 | XP_016864455.1 |
LARP1 - La ribonucleoprotein domain family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR378H - microRNA 378h | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |