Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAGAAAGCCCGAGCACTTCAC[A/G]TATCAAGAGGAAAACTATTGCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609014 | ||||||||||||||||||||
Literature Links: |
MCCC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCCC2 - methylcrotonoyl-CoA carboxylase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022132.4 | 364 | Missense Mutation | ATA,GTA | I,V 76 | NP_071415.1 | |
XM_005248567.1 | 364 | Missense Mutation | ATA,GTA | I,V 76 | XP_005248624.1 | |
XM_011543529.2 | 364 | Missense Mutation | ATA,GTA | I,V 76 | XP_011541831.1 | |
XM_017009688.1 | 364 | Missense Mutation | ATA,GTA | I,V 76 | XP_016865177.1 |