Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCGCACCCGCCGATTGTGGCCA[G/T]GGCGGCCGCAGTCTCTAGTGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf51 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf51 - chromosome 5 open reading frame 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175921.5 | 32 | Missense Mutation | AGG,ATG | R,M 1 | NP_787117.3 | |
XM_005248289.4 | 32 | Missense Mutation | AGG,ATG | R,M 1 | XP_005248346.1 | |
XM_011514032.2 | 32 | UTR 5 | XP_011512334.1 |