Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGGGCCGCACCTTTGCCGGACC[C/T]ATCCTCTGACTGCAGGAACGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142570 MIM: 607869 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HK3 - hexokinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002115.2 | 2796 | Missense Mutation | AGG,GGG | R,G 902 | NP_002106.2 | |
XM_011534540.2 | 2796 | Intron | XP_011532842.1 | |||
XM_017009411.1 | 2796 | Missense Mutation | AGG,GGG | R,G 845 | XP_016864900.1 |
UNC5A - unc-5 netrin receptor A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133369.2 | 2796 | Intron | NP_588610.2 | |||
XM_006714927.1 | 2796 | Intron | XP_006714990.1 | |||
XM_006714928.1 | 2796 | Intron | XP_006714991.1 | |||
XM_011534686.1 | 2796 | Intron | XP_011532988.1 | |||
XM_011534687.2 | 2796 | Intron | XP_011532989.1 |