Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGTTCTCCTTAAAGAAGTTAGG[G/T]GTAAACAATATCTCTGGTATTGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTF3 - basic transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037637.1 | 531 | Silent Mutation | GGG,GGT | G,G 96 | NP_001032726.1 | |
NM_001207.4 | 531 | Silent Mutation | GGG,GGT | G,G 52 | NP_001198.2 |