Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGGCCCCACGCAGCCACCAGA[C/T]AGCCATGCCCAGCAGCTTCCCACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616384 | ||||||||||||||||||||
Literature Links: |
UGT3A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGT3A2 - UDP glycosyltransferase family 3 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168316.1 | 1663 | Missense Mutation | ATC,GTC | I,V 477 | NP_001161788.1 | |
NM_174914.3 | 1663 | Missense Mutation | ATC,GTC | I,V 511 | NP_777574.2 | |
XM_011513988.2 | 1663 | Missense Mutation | ATC,GTC | I,V 538 | XP_011512290.1 | |
XM_017009150.1 | 1663 | Missense Mutation | ATC,GTC | I,V 473 | XP_016864639.1 |