Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGACGTATTTCCGCCGCTTGCTG[C/T]GGGTGGGCTGGCAGCACTGGGGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603745 | ||||||||||||||||||||
Literature Links: |
SLIT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLIT3 - slit guidance ligand 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271946.1 | 4892 | Missense Mutation | CAC,CGC | H,R 1498 | NP_001258875.1 | |
NM_003062.3 | 4892 | Missense Mutation | CAC,CGC | H,R 1491 | NP_003053.1 | |
XM_017009779.1 | 4892 | Missense Mutation | CAC,CGC | H,R 1428 | XP_016865268.1 |