Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCACTGCCCAGCCTACCTGTAG[C/T]CCCAGGGAGACAGGCTCCTCTTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604627 | ||||||||||||||||||||
Literature Links: |
IL17B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL17B - interleukin 17B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317987.1 | 678 | Missense Mutation | GAC,GGC | D,G 50 | NP_001304916.1 | |
NM_014443.2 | 678 | Missense Mutation | GAC,GGC | D,G 102 | NP_055258.1 | |
XM_011537622.2 | 678 | Missense Mutation | GAC,GGC | D,G 50 | XP_011535924.1 | |
XM_017009346.1 | 678 | Missense Mutation | GAC,GGC | D,G 167 | XP_016864835.1 | |
XM_017009347.1 | 678 | Missense Mutation | GAC,GGC | D,G 50 | XP_016864836.1 | |
XM_017009348.1 | 678 | Missense Mutation | GAC,GGC | D,G 50 | XP_016864837.1 |
PCYOX1L - prenylcysteine oxidase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |