Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCCCATTTCCAAAGCCTTTAG[G/T]GTATCTTCAACGCCAAAACAGTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600285 | ||||||||||||||||||||
Literature Links: |
ETF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETF1 - eukaryotic translation termination factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256302.1 | 1345 | Silent Mutation | NP_001243231.1 | |||
NM_001282185.1 | 1345 | Silent Mutation | NP_001269114.1 | |||
NM_001291974.1 | 1345 | Silent Mutation | NP_001278903.1 | |||
NM_001291975.1 | 1345 | Silent Mutation | NP_001278904.1 | |||
NM_004730.3 | 1345 | Silent Mutation | NP_004721.1 | |||
XM_005271921.1 | 1345 | Silent Mutation | XP_005271978.1 | |||
XM_017009221.1 | 1345 | Intron | XP_016864710.1 | |||
XM_017009222.1 | 1345 | Silent Mutation | XP_016864711.1 |