Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCATTGCTGATTTCCTTCTGAAA[C/G]ATAAGATTGACAATTTATTTCATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD34B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD34B - ankyrin repeat domain 34B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004441.2 | 1412 | Missense Mutation | CTT,GTT | L,V 328 | NP_001004441.2 | |
XM_006714599.3 | 1412 | Missense Mutation | CTT,GTT | L,V 328 | XP_006714662.1 | |
XM_011543366.2 | 1412 | Missense Mutation | CTT,GTT | L,V 328 | XP_011541668.1 | |
XM_011543369.2 | 1412 | Missense Mutation | CTT,GTT | L,V 328 | XP_011541671.1 |