Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCATTTTGGGCATTTCAGCTCAG[C/T]TGTCATCTCAGTGACCAGTTTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608039 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFESD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFESD - Rieske Fe-S domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131065.1 | 1347 | Intron | NP_001124537.1 | |||
NM_001131066.1 | 1347 | Intron | NP_001124538.1 | |||
NM_173362.3 | 1347 | Intron | NP_775498.1 | |||
XM_011543360.2 | 1347 | Missense Mutation | GCT,GTT | A,V 43 | XP_011541662.1 | |
XM_011543361.2 | 1347 | Intron | XP_011541663.1 | |||
XM_017009413.1 | 1347 | Missense Mutation | GCT,GTT | A,V 43 | XP_016864902.1 |
SPATA9 - spermatogenesis associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031952.3 | 1347 | Intron | NP_114158.2 | |||
XM_011543664.1 | 1347 | Intron | XP_011541966.1 | |||
XM_011543666.1 | 1347 | Intron | XP_011541968.1 | |||
XM_011543667.2 | 1347 | Intron | XP_011541969.1 | |||
XM_017009948.1 | 1347 | Intron | XP_016865437.1 | |||
XM_017009949.1 | 1347 | Intron | XP_016865438.1 | |||
XM_017009950.1 | 1347 | Intron | XP_016865439.1 | |||
XM_017009951.1 | 1347 | Intron | XP_016865440.1 |