Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGTCAGAACCATGGACAAAACC[A/G]TGTTGGCATCAATGAGATATGATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616704 MIM: 605885 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COMMD10 - COMM domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308080.1 | 4971 | Intron | NP_001295009.1 | |||
NM_016144.3 | 4971 | Intron | NP_057228.1 | |||
XM_011543458.2 | 4971 | Intron | XP_011541760.1 | |||
XM_011543460.2 | 4971 | Intron | XP_011541762.1 | |||
XM_017009553.1 | 4971 | Intron | XP_016865042.1 | |||
XM_017009555.1 | 4971 | Intron | XP_016865044.1 | |||
XM_017009556.1 | 4971 | Intron | XP_016865045.1 | |||
XM_017009557.1 | 4971 | Intron | XP_016865046.1 |
SEMA6A - semaphorin 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300780.1 | 4971 | UTR 3 | NP_001287709.1 | |||
NM_020796.4 | 4971 | UTR 3 | NP_065847.1 | |||
XM_005272042.3 | 4971 | UTR 3 | XP_005272099.2 | |||
XM_006714663.1 | 4971 | UTR 3 | XP_006714726.1 | |||
XM_017009675.1 | 4971 | UTR 3 | XP_016865164.1 | |||
XM_017009676.1 | 4971 | UTR 3 | XP_016865165.1 |
SEMA6A-AS1 - SEMA6A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |