Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACTGCTGGACCAGAATGACCCT[A/G]TAATTTCCTAAGATCATTCTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE8B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE8B - phosphodiesterase 8B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029851.2 | Intron | NP_001025022.1 | ||||
NM_001029852.2 | Intron | NP_001025023.1 | ||||
NM_001029853.2 | Intron | NP_001025024.1 | ||||
NM_001029854.2 | Intron | NP_001025025.1 | ||||
NM_003719.3 | Intron | NP_003710.1 | ||||
XM_005248621.4 | Intron | XP_005248678.1 | ||||
XM_005248623.4 | Intron | XP_005248680.1 | ||||
XM_005248624.4 | Intron | XP_005248681.1 | ||||
XM_006714725.3 | Intron | XP_006714788.1 | ||||
XM_006714726.3 | Intron | XP_006714789.1 | ||||
XM_011543699.2 | Intron | XP_011542001.1 | ||||
XM_011543700.2 | Intron | XP_011542002.1 | ||||
XM_011543701.2 | Intron | XP_011542003.1 | ||||
XM_011543702.2 | Intron | XP_011542004.1 | ||||
XM_011543703.2 | Intron | XP_011542005.1 | ||||
XM_011543704.2 | Intron | XP_011542006.1 | ||||
XM_017010003.1 | Intron | XP_016865492.1 | ||||
XM_017010004.1 | Intron | XP_016865493.1 | ||||
XM_017010005.1 | Intron | XP_016865494.1 | ||||
XM_017010006.1 | Intron | XP_016865495.1 | ||||
XM_017010007.1 | Intron | XP_016865496.1 | ||||
XM_017010008.1 | Intron | XP_016865497.1 |
WDR41 - WD repeat domain 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018268.3 | Intron | NP_060738.2 | ||||
XM_005248551.4 | Intron | XP_005248608.1 | ||||
XM_005248552.3 | Intron | XP_005248609.1 | ||||
XM_011543505.2 | Intron | XP_011541807.1 | ||||
XM_011543507.2 | Intron | XP_011541809.1 | ||||
XM_017009606.1 | Intron | XP_016865095.1 |