Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTCGCACTGTGAGGATATTCT[C/T]TGGGGTCAAAAGCCCCTTCCTGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RNF145 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF145 - ring finger protein 145 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199380.1 | 2153 | Missense Mutation | AAA,AGA | K,R 677 | NP_001186309.1 | |
NM_001199381.1 | 2153 | Missense Mutation | AAA,AGA | K,R 664 | NP_001186310.1 | |
NM_001199382.1 | 2153 | Missense Mutation | AAA,AGA | K,R 661 | NP_001186311.1 | |
NM_001199383.1 | 2153 | Missense Mutation | AAA,AGA | K,R 647 | NP_001186312.1 | |
NM_144726.2 | 2153 | Missense Mutation | AAA,AGA | K,R 675 | NP_653327.1 | |
XM_017009138.1 | 2153 | Missense Mutation | AAA,AGA | K,R 647 | XP_016864627.1 |