Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCACCTCCTCCTCCACAGGTGC[A/G]GCCGACCAGGTGGACCCTGGCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126060 MIM: 600887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHFR - dihydrofolate reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000791.3 | 209 | UTR 5 | NP_000782.1 | |||
NM_001290354.1 | 209 | UTR 5 | NP_001277283.1 | |||
NM_001290357.1 | 209 | UTR 5 | NP_001277286.1 |
MSH3 - mutS homolog 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002439.4 | 209 | Silent Mutation | GCA,GCG | A,A 43 | NP_002430.3 |
MTRNR2L2 - MT-RNR2-like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |