Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAATTGAAGGCAACGGTAAAAG[A/G]AATGTATAGGCCAGGACGTGAACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT5 - chaperonin containing TCP1 subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306153.1 | Intron | NP_001293082.1 | ||||
NM_001306154.1 | Intron | NP_001293083.1 | ||||
NM_001306155.1 | Intron | NP_001293084.1 | ||||
NM_001306156.1 | Intron | NP_001293085.1 | ||||
NM_012073.4 | Intron | NP_036205.1 |
FAM173B - family with sequence similarity 173 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |