Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATCCATTTTTCACCTTATGGTG[A/C]ATTTAGTATACAATTATCCATATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGCS1 - 3-hydroxy-3-methylglutaryl-CoA synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098272.2 | Intron | NP_001091742.1 | ||||
NM_001324219.1 | Intron | NP_001311148.1 | ||||
NM_001324220.1 | Intron | NP_001311149.1 | ||||
NM_001324222.1 | Intron | NP_001311151.1 | ||||
NM_001324223.1 | Intron | NP_001311152.1 | ||||
NM_001324224.1 | Intron | NP_001311153.1 | ||||
NM_002130.7 | Intron | NP_002121.4 | ||||
XM_011514036.2 | Intron | XP_011512338.1 | ||||
XM_017009412.1 | Intron | XP_016864901.1 |