Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGACCGGTGGTCTGAATGTAACC[A/G]ATGATCTGAGTGAGAACGATGATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602118 | ||||||||||||||||||||
Literature Links: |
CHD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHD1 - chromodomain helicase DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270.2 | 2860 | Missense Mutation | CGG,TGG | R,W 1638 | NP_001261.2 | |
XM_005271866.4 | 2860 | Missense Mutation | CGG,TGG | R,W 1726 | XP_005271923.1 | |
XM_005271867.4 | 2860 | Missense Mutation | CGG,TGG | R,W 1638 | XP_005271924.1 | |
XM_011543112.2 | 2860 | UTR 3 | XP_011541414.1 | |||
XM_017008991.1 | 2860 | Missense Mutation | CGG,TGG | R,W 797 | XP_016864480.1 | |
XM_017008992.1 | 2860 | Missense Mutation | CGG,TGG | R,W 797 | XP_016864481.1 |