Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCTGGGGCTCTGGCTCTGGTTC[A/G]TCCTGGGGCTCCAGGGGGCTGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164770 MIM: 173410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSF1R - colony stimulating factor 1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDGFRB - platelet derived growth factor receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002609.3 | 4153 | Silent Mutation | GAC,GAT | D,D 1068 | NP_002600.1 | |
XM_005268464.2 | 4153 | Silent Mutation | GAC,GAT | D,D 1004 | XP_005268521.1 | |
XM_011537658.1 | 4153 | Silent Mutation | GAC,GAT | D,D 1068 | XP_011535960.1 | |
XM_011537659.1 | 4153 | Silent Mutation | GAC,GAT | D,D 1068 | XP_011535961.1 |