Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACCACTAGACAGAGAAAGCAGA[A/G]CCGAGTACAACGTCACTATCACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606345 MIM: 606333 MIM: 606334 MIM: 606335 | ||||||||||||||||||||
Literature Links: |
PCDHB16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDHB16 - protocadherin beta 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB7 - protocadherin beta 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB8 - protocadherin beta 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019120.4 | 1491 | Missense Mutation | ACC,GCC | T,A 416 | NP_061993.3 |
PCDHB9 - protocadherin beta 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |