Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAAAAGCCATTTTCTTCTGGGA[A/G]AACTATTACCAAACACCAGGTAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607819 | ||||||||||||||||||||
Literature Links: |
SLC30A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC30A5 - solute carrier family 30 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251969.1 | 391 | Missense Mutation | AAA,AGA | K,R 44 | NP_001238898.1 | |
NM_022902.4 | 391 | Missense Mutation | AAA,AGA | K,R 85 | NP_075053.2 | |
NM_024055.4 | 391 | Missense Mutation | AAA,AGA | K,R 85 | NP_076960.1 | |
XM_005248569.3 | 391 | Missense Mutation | AAA,AGA | K,R 44 | XP_005248626.1 | |
XM_006714672.3 | 391 | Missense Mutation | AAA,AGA | K,R 85 | XP_006714735.1 | |
XM_017009749.1 | 391 | Missense Mutation | AAA,AGA | K,R 44 | XP_016865238.1 |