Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGACCTCGCTGCCAGAACGGGGT[C/T]GCTTTGACAAGGTGCACCTTTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
N4BP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
N4BP3 - NEDD4 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015111.1 | 566 | Missense Mutation | CGC,TGC | R,C 107 | NP_055926.1 | |
XM_006714834.3 | 566 | Missense Mutation | CGC,TGC | R,C 107 | XP_006714897.1 | |
XM_011534473.1 | 566 | Missense Mutation | CGC,TGC | R,C 107 | XP_011532775.1 | |
XM_011534474.1 | 566 | Missense Mutation | CGC,TGC | R,C 107 | XP_011532776.1 |