Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAAAGAAGAATCTCAACCGA[A/T]CTGGTAAGCGTGGCCGGCCTTCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C5orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5orf24 - chromosome 5 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135586.1 | 675 | Missense Mutation | ACT,TCT | T,S 87 | NP_001129058.1 | |
NM_001300894.1 | 675 | Missense Mutation | ACT,TCT | T,S 87 | NP_001287823.1 | |
NM_152409.3 | 675 | Missense Mutation | ACT,TCT | T,S 87 | NP_689622.2 | |
XM_005271889.3 | 675 | Missense Mutation | ACT,TCT | T,S 87 | XP_005271946.1 | |
XM_017009049.1 | 675 | Missense Mutation | ACT,TCT | T,S 87 | XP_016864538.1 | |
XM_017009050.1 | 675 | Missense Mutation | ACT,TCT | T,S 87 | XP_016864539.1 |