Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGCTGCGCAGCCCCAGGCCTG[A/T]ATCGATCACCACGTCCTGCTCGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBLAC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBLAC2 - metallo-beta-lactamase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203406.1 | 229 | Missense Mutation | ACA,TCA | T,S 46 | NP_981951.1 |
POLR3G - RNA polymerase III subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006467.2 | 229 | Intron | NP_006458.2 | |||
XM_005248404.3 | 229 | Intron | XP_005248461.1 | |||
XM_011543100.2 | 229 | UTR 5 | XP_011541402.1 | |||
XM_011543101.2 | 229 | Intron | XP_011541403.1 | |||
XM_011543102.2 | 229 | Intron | XP_011541404.1 | |||
XM_011543103.2 | 229 | Intron | XP_011541405.1 | |||
XM_011543104.2 | 229 | Intron | XP_011541406.1 | |||
XM_011543105.2 | 229 | Intron | XP_011541407.1 | |||
XM_017008957.1 | 229 | Intron | XP_016864446.1 |