Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCAGATTCGCAAGTAGGTGCCC[A/G]TGGAGCAAACAGAGAAAAGCGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102577426 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102577426 - uncharacterized LOC102577426 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM7 - tripartite motif containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033342.3 | 1582 | Intron | NP_203128.1 | |||
NM_203293.2 | 1582 | Missense Mutation | ACG,ATG | T,M 503 | NP_976038.1 | |
NM_203294.1 | 1582 | Missense Mutation | ACG,ATG | T,M 295 | NP_976039.1 | |
NM_203295.1 | 1582 | Missense Mutation | ACG,ATG | T,M 295 | NP_976040.1 | |
NM_203296.1 | 1582 | Missense Mutation | ACG,ATG | T,M 295 | NP_976041.1 | |
NM_203297.1 | 1582 | Missense Mutation | ACG,ATG | T,M 321 | NP_976042.1 | |
XM_017009903.1 | 1582 | Missense Mutation | ACG,ATG | T,M 295 | XP_016865392.1 | |
XM_017009904.1 | 1582 | Intron | XP_016865393.1 |