Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCAAAATGAATTACAGAATCAA[C/T]TGGATAAATGTCGTAATGAAATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616401 | ||||||||||||||||||||
Literature Links: |
SPDL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPDL1 - spindle apparatus coiled-coil protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017785.4 | 295 | Silent Mutation | CTG,TTG | L,L 41 | NP_060255.3 | |
XM_011534585.1 | 295 | Silent Mutation | CTG,TTG | L,L 41 | XP_011532887.1 | |
XM_011534586.2 | 295 | Silent Mutation | CTG,TTG | L,L 41 | XP_011532888.1 | |
XM_011534587.2 | 295 | Silent Mutation | CTG,TTG | L,L 41 | XP_011532889.1 | |
XM_011534588.2 | 295 | Silent Mutation | CTG,TTG | L,L 41 | XP_011532890.1 | |
XM_011534589.2 | 295 | Silent Mutation | CTG,TTG | L,L 41 | XP_011532891.1 | |
XM_011534590.2 | 295 | Intron | XP_011532892.1 | |||
XM_017009601.1 | 295 | Silent Mutation | CTG,TTG | L,L 41 | XP_016865090.1 | |
XM_017009602.1 | 295 | Intron | XP_016865091.1 |