Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACTTTGTCAAGGCTTCTTAGAA[C/T]GTGTTGAAATAAGGTCAAAGCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102467212 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102467212 - uncharacterized LOC102467212 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT12 - nudix hydrolase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300741.1 | 2423 | UTR 3 | NP_001287670.1 | |||
NM_031438.3 | 2423 | UTR 3 | NP_113626.1 | |||
XM_005272095.1 | 2423 | UTR 3 | XP_005272152.1 | |||
XM_005272097.3 | 2423 | UTR 3 | XP_005272154.1 |