Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGAGCTGTTCTGGGAATTCCTAT[C/T]GCTGTGCCTCTGAACTGAGGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601893 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIO - trio Rho guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007118.3 | Intron | NP_009049.2 | ||||
XM_011514107.2 | Intron | XP_011512409.1 | ||||
XM_011514108.1 | Intron | XP_011512410.1 | ||||
XM_011514109.2 | Intron | XP_011512411.1 | ||||
XM_011514110.2 | Intron | XP_011512412.1 | ||||
XM_017009801.1 | Intron | XP_016865290.1 | ||||
XM_017009802.1 | Intron | XP_016865291.1 | ||||
XM_017009803.1 | Intron | XP_016865292.1 |