Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGAGAAGCTGGACCGGGAGAAG[A/C]TGTGTGGCCCTACTGAGCCCTGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606341 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDHB15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDHB15 - protocadherin beta 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018935.3 | 513 | Missense Mutation | ATG,CTG | M,L 95 | NP_061758.1 |
PCDHB18P - protocadherin beta 18 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDHB19P - protocadherin beta 19 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |