Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGAAATAAAGTCAAGTGCCAG[C/T]GGTCTCTGCACGCTGCTCCAGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDNF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDNF - glial cell derived neurotrophic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000514.3 | 2132 | UTR 3 | NP_000505.1 | |||
NM_001190468.1 | 2132 | UTR 3 | NP_001177397.1 | |||
NM_001190469.1 | 2132 | UTR 3 | NP_001177398.1 | |||
NM_001278098.1 | 2132 | UTR 3 | NP_001265027.1 | |||
NM_199231.2 | 2132 | UTR 3 | NP_954701.1 | |||
XM_011514030.2 | 2132 | Intron | XP_011512332.1 | |||
XM_017009337.1 | 2132 | Intron | XP_016864826.1 |