Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCTGCCTTTTGTTTCATTGCCG[C/T]GTTGGTGATCTTTGTTACCAGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602876 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCLN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OCLN - occludin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205254.1 | 891 | Missense Mutation | GCG,GTG | A,V 152 | NP_001192183.1 | |
NM_001205255.1 | 891 | Intron | NP_001192184.1 | |||
NM_002538.3 | 891 | Missense Mutation | GCG,GTG | A,V 152 | NP_002529.1 | |
XM_017008913.1 | 891 | Missense Mutation | GCG,GTG | A,V 152 | XP_016864402.1 | |
XM_017008914.1 | 891 | Missense Mutation | GCG,GTG | A,V 152 | XP_016864403.1 |