Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTATGGGGAGTCTCGTGAGGCC[A/G]GCACCAAAGAGGATTAGGCTACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAIP - NLR family apoptosis inhibitory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004536.2 | 1039 | Silent Mutation | NP_004527.2 | |||
NM_022892.1 | 1039 | Intron | NP_075043.1 | |||
XM_011543413.2 | 1039 | Intron | XP_011541715.1 | |||
XM_017009490.1 | 1039 | Intron | XP_016864979.1 |