Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCTGGCTGCGCTCGGCCTGCTC[A/G]CCCACTTCCTCCCGGCCGCGCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 246530 MIM: 604561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTC4S PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LTC4S - leukotriene C4 synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145867.1 | 478 | Missense Mutation | ACC,GCC | T,A 128 | NP_665874.1 |
MGAT4B - mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1229 - microRNA 1229 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |