Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTTCTGTGCCACGAGGCCGGAG[C/T]TGAGCCGCTGCGTCGCCTTCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600869 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRK6 - G protein-coupled receptor kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004105.2 | 389 | Silent Mutation | CTG,TTG | L,L 77 | NP_001004105.1 | |
NM_001004106.2 | 389 | Silent Mutation | CTG,TTG | L,L 77 | NP_001004106.1 | |
NM_002082.3 | 389 | Silent Mutation | CTG,TTG | L,L 77 | NP_002073.2 | |
XM_006714858.3 | 389 | Silent Mutation | CTG,TTG | L,L 47 | XP_006714921.1 | |
XM_006714859.2 | 389 | Silent Mutation | CTG,TTG | L,L 45 | XP_006714922.1 | |
XM_011534537.2 | 389 | Intron | XP_011532839.1 | |||
XM_011534538.1 | 389 | Intron | XP_011532840.1 |
PRR7-AS1 - PRR7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |